2-Fluorophenyl isopropylcarbamate ≥95%

Supplier: AOBChem USA

Warning

16729-500MG 16729-1G 16729-25G 16729-5G
77324-098EA 289.43 USD
77324-098 77348-080 77383-900 77372-286
2-Fluorophenyl isopropylcarbamate ≥95%
2-Fluorophenyl isopropylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034166
CAS Number: 199585-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR