2-(Benzyloxy)-6-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

16728-500MG 16728-1G 16728-5G 16728-25G
77324-096EA 202.91 USD
77324-096 77348-078 77360-340 77383-898
2-(Benzyloxy)-6-fluoro-N-methoxy-N-methylbenzamide ≥97%
2-(Benzyloxy)-6-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₆H₁₆FNO₃
MW: 289.302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755702
CAS Number: 2901096-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR