2-Fluoro-6-hydroxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16653-500MG 16653-1G 16653-5G 16653-25G
77324-094EA 206 USD
77324-094 77348-076 77360-338 77383-896
2-Fluoro-6-hydroxy-3-methoxybenzaldehyde ≥97%
2-Fluoro-6-hydroxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520915
CAS Number: 783342-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR