2,3-Difluoro-6-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16651-500MG 16651-1G 16651-5G 16651-25G
77324-092EA 222.48 USD
77324-092 77348-074 77360-336 77383-894
2,3-Difluoro-6-isopropoxybenzaldehyde ≥95%
2,3-Difluoro-6-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27934421
CAS Number: 949026-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR