Methyl 2,5-difluoro-3-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

16649-500MG 16649-1G 16649-5G 16649-25G
77324-090EA 202.91 USD
77324-090 77348-072 77360-334 77383-892
Methyl 2,5-difluoro-3-formylbenzoate ≥97%
Methyl 2,5-difluoro-3-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977397
CAS Number: 952480-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR