2-Iodothiophene-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

166463-500MG 166463-1G 166463-5G 166463-25G
77324-088EA 271.92 USD
77324-088 77348-070 77360-332 77383-890
2-Iodothiophene-3-carboxylic acid ≥95%
2-Iodothiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃IO₂S
Storage Temperature: Ambient
CAS Number: 18895-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR