3-(4-Bromophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166184-500MG 166184-1G 166184-5G 166184-25G
77324-086EA 93.73 USD
77324-086 77342-518 77360-330 77372-284
3-(4-Bromophenyl)oxetan-3-ol ≥95%
3-(4-Bromophenyl)oxetan-3-ol

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16657878
CAS Number: 1093878-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR