2,4-Dichloro-5-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

16479-500MG 16479-1G 16479-5G 16479-25G
77324-082EA 204.97 USD
77324-082 77348-066 77360-322 77383-886
2,4-Dichloro-5-(trifluoromethyl)aniline ≥97%
2,4-Dichloro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃N
MW: 230.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828386
CAS Number: 320-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR