1,2-Dibenzyloxybenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Benzylcyclohexa-2,4-dienone

16366-500MG 16366-1G 16366-5G 16366-25G
77324-074EA 215.27 USD
77324-074 77348-058 77360-312 77383-882
1,2-Dibenzyloxybenzene ≥97%
1,2-Dibenzyloxybenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₂
MW: 290.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00401895
CAS Number: 10403-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR