2,4-Dichloro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16307-500MG 16307-1G 16307-5G 16307-25G
77324-072EA 141.11 USD
77324-072 77344-770 77360-310 77383-880
2,4-Dichloro-3-formylphenylboronic acid ≥97%
2,4-Dichloro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BCl₂O₃
MW: 218.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380417
CAS Number: 480438-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR