2,6-Dichloro-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16305-1G 16305-5G 16305-25G
77341-588EA 94.76 USD
77341-588 77360-308 77372-264
2,6-Dichloro-4-(methylthio)benzaldehyde ≥97%
2,6-Dichloro-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27926360
CAS Number: 616196-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR