3-(Benzyloxy)-2-chloro-6-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
16271-500MG
16271-1G
16271-5G
16271-25G
77324-068EA
144.2
USD
77324-068
77345-032
77360-304
77383-876
3-(Benzyloxy)-2-chloro-6-fluorobenzaldehyde ≥97%
3-(Benzyloxy)-2-chloro-6-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₀ClFO₂ MW: 264.679 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520665 CAS Number: 918524-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...