N-(2,4-Dibromophenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

162231-500MG 162231-1G 162231-5G
77337-032EA 405.82 USD
77337-032 77348-056 77372-258
N-(2,4-Dibromophenyl)acetamide ≥97%
N-(2,4-Dibromophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO
MW: 292.9552 g/mol
Storage Temperature: Ambient
CAS Number: 23373-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR