2-Chloro-4-fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16221-500MG 16221-1G 16221-5G
77336-230EA 384.19 USD
77336-230 77348-054 77372-256
2-Chloro-4-fluoro-3-hydroxybenzaldehyde ≥97%
2-Chloro-4-fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18824352
CAS Number: 960001-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR