2,5-Difluoro-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16168-500MG 16168-1G 16168-5G 16168-25G
77324-066EA 254.41 USD
77324-066 77348-052 77360-298 77383-874
2,5-Difluoro-4-methylphenylboronic acid ≥97%
2,5-Difluoro-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20485030
CAS Number: 872219-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR