3-Phenyloxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

161589-500MG 161589-1G 161589-5G 161589-25G
77324-064EA 182.31 USD
77324-064 77347-108 77360-296 77383-872
3-Phenyloxetan-3-ol ≥95%
3-Phenyloxetan-3-ol

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD18909281
CAS Number: 699-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR