Ethyl(4-fluoro-2-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161547-500MG 161547-1G 161547-5G 161547-25G
77324-062EA 243.08 USD
77324-062 77348-050 77360-294 77383-870
Ethyl(4-fluoro-2-methylphenyl)sulfane ≥95%
Ethyl(4-fluoro-2-methylphenyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FS
Storage Temperature: Ambient
CAS Number: 1314976-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR