3-Bromo-4-ethylaniline ≥97%

Supplier: AOBChem USA

Warning

161032-500MG 161032-1G 161032-5G 161032-25G
77324-058EA 104.03 USD
77324-058 77343-084 77360-288 77372-248
3-Bromo-4-ethylaniline ≥97%
3-Bromo-4-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.0757 g/mol
Storage Temperature: Ambient
CAS Number: 52121-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR