2-Bromo-5-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16073-500MG 16073-1G 16073-5G
77340-084EA 753.96 USD
77340-084 77348-042 77372-244
2-Bromo-5-formylphenylboronic acid ≥97%
2-Bromo-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₃
MW: 228.836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28167204
CAS Number: 1008106-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR