3,4-Dichloro-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16058-500MG 16058-1G 16058-5G
77338-030EA 460.41 USD
77338-030 77348-040 77372-242
3,4-Dichloro-5-fluorophenylboronic acid ≥97%
3,4-Dichloro-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD23380982
CAS Number: 1023690-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR