(3-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

16054-500MG 16054-1G 16054-5G 16054-25G
77324-054EA 86.52 USD
77324-054 77342-236 77360-282 77372-240
(3-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥95%
(3-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609514
CAS Number: 1026796-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR