2,6-Diethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

16038-1G 16038-5G 16038-25G
77340-588EA 60.77 USD
77340-588 77360-280 77372-238
2,6-Diethoxypyridine ≥97%
2,6-Diethoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234572
CAS Number: 13472-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR