2-Bromo-6-chloro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15971-500MG 15971-1G 15971-5G 15971-25G
77324-052EA 152.44 USD
77324-052 77345-442 77360-278 77383-864
2-Bromo-6-chloro-3-methoxybenzaldehyde ≥97%
2-Bromo-6-chloro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD30829702
CAS Number: 2089650-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR