1,4-Dibromo-2-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15958-500MG 15958-1G 15958-5G 15958-25G
77324-050EA 244.11 USD
77324-050 77348-036 77360-276 77383-862
1,4-Dibromo-2-fluoro-3-methoxybenzene ≥97%
1,4-Dibromo-2-fluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28784584
CAS Number: 919355-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR