2-Methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

15877-500MG 15877-1G 15877-5G 15877-25G
77324-046EA 204.97 USD
77324-046 77348-032 77360-272 77383-860
2-Methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥95%
2-Methoxy-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O
MW: 252.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702105
CAS Number: 1214380-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR