1,4-Difluoro-2,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15869-500MG 15869-1G 15869-5G
77324-044EA 353.29 USD
77324-044 77348-030 77372-230
1,4-Difluoro-2,3-dimethoxybenzene ≥97%
1,4-Difluoro-2,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394834
CAS Number: 75626-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR