5-(Ethylthio)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

158475-500MG 158475-1G 158475-5G 158475-25G
77324-042EA 243.08 USD
77324-042 77348-028 77360-270 77383-858
5-(Ethylthio)benzo[d][1,3]dioxole ≥97%
5-(Ethylthio)benzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
CAS Number: 5279-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR