5-(Methylthio)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

158473-500MG 158473-1G 158473-5G 158473-25G
77324-040EA 186.43 USD
77324-040 77348-026 77360-268 77383-856
5-(Methylthio)benzo[d][1,3]dioxole ≥97%
5-(Methylthio)benzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₈H₈O₂S
MW: 168.213 g/mol
Storage Temperature: Ambient
CAS Number: 5354-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR