2,3-Difluoro-6-(phenylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15803-500MG 15803-1G 15803-5G 15803-25G
77324-036EA 113.3 USD
77324-036 77343-340 77360-264 77372-224
2,3-Difluoro-6-(phenylmethoxy)benzaldehyde ≥97%
2,3-Difluoro-6-(phenylmethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₂
MW: 248.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520663
CAS Number: 947279-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR