1-Chloro-4-fluoro-2-iodo-5-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
15792-500MG
15792-1G
15792-5G
77335-490EA
356.38
USD
77335-490
77348-022
77372-222
1-Chloro-4-fluoro-2-iodo-5-methoxybenzene ≥97%
1-Chloro-4-fluoro-2-iodo-5-methoxybenzene
Precaución: for Research Use Only
Formula:
C₇H₅ClFIO MW: 286.47 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28976839 CAS Number: 677741-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...