3,5-Dibromo-4-butoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15731-500MG 15731-1G 15731-5G 15731-25G
77324-034EA 119.48 USD
77324-034 77343-626 77360-258 77372-216
3,5-Dibromo-4-butoxybenzaldehyde ≥95%
3,5-Dibromo-4-butoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12030622
CAS Number: 1134336-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR