3-Bromo-5-chloro-4-(phenylmethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15724-500MG 15724-1G 15724-5G 15724-25G
77324-032EA 110.21 USD
77324-032 77343-252 77360-256 77372-214
3-Bromo-5-chloro-4-(phenylmethoxy)benzaldehyde ≥95%
3-Bromo-5-chloro-4-(phenylmethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrClO₂
MW: 325.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02629408
CAS Number: 872136-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR