4-Bromo-2,6-difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

15665-500MG 15665-1G 15665-5G 15665-25G
77324-028EA 181.28 USD
77324-028 77346-772 77360-250 77383-852
4-Bromo-2,6-difluoro-N-methoxy-N-methylbenzamide ≥97%
4-Bromo-2,6-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD24801939
CAS Number: 1449008-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR