2,4-Dibromo-1-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15652-500MG 15652-1G 15652-5G 15652-25G
77324-026EA 177.16 USD
77324-026 77346-574 77360-248 77383-850
2,4-Dibromo-1-ethoxybenzene ≥97%
2,4-Dibromo-1-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD11639121
CAS Number: 38751-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR