1-Fluoro-3-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15599-500MG 15599-1G 15599-5G 15599-25G
77324-024EA 120.51 USD
77324-024 77343-660 77360-246 77372-208
1-Fluoro-3-propoxybenzene ≥97%
1-Fluoro-3-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD01861234
CAS Number: 203115-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR