2-(2-Bromo-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15556-1G 15556-5G 15556-25G
77341-660EA 98.88 USD
77341-660 77360-242 77372-204
2-(2-Bromo-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-5-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976820
CAS Number: 1394824-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR