2-Bromo-1-ethoxy-4-trifluoromethyl-benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
15541-500MG
15541-1G
15541-5G
15541-25G
77324-022EA
211.15
USD
77324-022
77348-016
77360-240
77383-848
2-Bromo-1-ethoxy-4-trifluoromethyl-benzene ≥97%
2-Bromo-1-ethoxy-4-trifluoromethyl-benzene
Precaución: for Research Use Only
Formula:
C₉H₈BrF₃O MW: 269.058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11036038 CAS Number: 934495-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...