2-Bromo-1-isopropoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

15540-500MG 15540-1G 15540-5G 15540-25G
77324-020EA 211.15 USD
77324-020 77348-014 77360-238 77383-846
2-Bromo-1-isopropoxy-4-(trifluoromethyl)benzene ≥95%
2-Bromo-1-isopropoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570347
CAS Number: 200956-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR