2,4,6-Trifluoro-1-n-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15472-1G 15472-5G 15472-25G
77340-984EA 66.95 USD
77340-984 77360-234 77372-200
2,4,6-Trifluoro-1-n-propoxybenzene ≥97%
2,4,6-Trifluoro-1-n-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD19105350
CAS Number: 203511-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR