3-Bromo-4-(dimethylamino)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15388-1G 15388-5G 15388-25G
77340-586EA 60.77 USD
77340-586 77360-232 77372-198
3-Bromo-4-(dimethylamino)benzaldehyde ≥95%
3-Bromo-4-(dimethylamino)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD01593654
CAS Number: 56479-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR