3-Borono-4,5-dichlorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

15282-1G 15282-5G 15282-25G
77341-496EA 90.64 USD
77341-496 77360-222 77372-190
3-Borono-4,5-dichlorobenzoic acid ≥95%
3-Borono-4,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BCl₂O₄
Storage Temperature: Ambient
MDL Number: MFCD29035380
CAS Number: 2377608-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR