(2,3-Dichloro-5-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

15281-500MG 15281-1G 15281-5G 15281-25G
77324-008EA 191.58 USD
77324-008 77348-004 77360-220 77383-836
(2,3-Dichloro-5-methylphenyl)boronic acid ≥95%
(2,3-Dichloro-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₂
MW: 204.846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787023
CAS Number: 2377606-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR