Ethyl 4-bromo-3-chloro-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

15268-500MG 15268-1G 15268-5G
77337-868EA 450.11 USD
77337-868 77348-000 77372-188
Ethyl 4-bromo-3-chloro-2-methylbenzoate ≥95%
Ethyl 4-bromo-3-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32879594
CAS Number: 2384581-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR