1-(2-Fluoropyridin-3-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

152387-500MG 152387-1G 152387-5G
77324-002EA 347.11 USD
77324-002 77347-996 77372-186
1-(2-Fluoropyridin-3-yl)cyclohexanol ≥97%
1-(2-Fluoropyridin-3-yl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO
MW: 195.233 g/mol
Storage Temperature: Ambient
CAS Number: 2921797-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR