2-Chloro-4-fluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15238-500MG 15238-1G 15238-5G
77336-446EA 388.31 USD
77336-446 77347-994 77372-184
2-Chloro-4-fluoro-5-methoxybenzaldehyde ≥97%
2-Chloro-4-fluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297304
CAS Number: 177034-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR