5-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

151332-500MG 151332-1G 151332-5G 151332-25G
77323-998EA 246.17 USD
77323-998 77347-986 77360-212 77383-830
5-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide ≥97%
5-Bromo-2,3-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
Storage Temperature: Ambient
CAS Number: 1600511-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR