2-Bromo-6-chloro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15069-1G 15069-5G 15069-25G
77340-884EA 62.83 USD
77340-884 77360-206 77372-170
2-Bromo-6-chloro-4-methylbenzaldehyde ≥97%
2-Bromo-6-chloro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755670
CAS Number: 2090975-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR