(2-Cyano-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

15064-500MG 15064-1G 15064-5G 15064-25G
77323-992EA 177.16 USD
77323-992 77346-572 77360-204 77383-828
(2-Cyano-5-methylphenyl)boronic acid ≥97%
(2-Cyano-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394415
CAS Number: 1375109-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR