1,3-Dibromo-2,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

150315-500MG 150315-1G 150315-5G 150315-25G
77323-988EA 161.71 USD
77323-988 77346-072 77360-200 77383-826
1,3-Dibromo-2,5-dimethoxybenzene ≥95%
1,3-Dibromo-2,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
Storage Temperature: Ambient
CAS Number: 74076-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR