2-Bromo-5-butoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

14984-500MG 14984-1G 14984-5G 14984-25G
77323-986EA 87.55 USD
77323-986 77342-278 77360-196 77372-158
2-Bromo-5-butoxy-1,3-dimethylbenzene ≥97%
2-Bromo-5-butoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
MW: 257.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917837
CAS Number: 91560-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR